N,N,N',N'-Tetrakis(p-tolyl)benzidine ≥98.0% (by HPLC, total nitrogen), purified by sublimation

Supplier: TCI America

Synonyms: N4,N4,N4',N4'-Tetra-p-tolyl-[1,1'-biphenyl]-4,4'-diamine, N,N,N',N'-Tetrakis(4-methylphenyl)benzidine

T3265-1G
TCT3265-1GEA 113 USD
TCT3265-1G
N,N,N',N'-Tetrakis(p-tolyl)benzidine ≥98.0% (by HPLC, total nitrogen), purified by sublimation
N,N,N',N'-Tetrakis(p-tolyl)benzidine
Formula: C₄₀H₃₆N₂
MW: 544.74 g/mol
MDL Number: MFCD00799300
CAS Number: 76185-65-4

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %
Melting point 216.0 to 220.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR