Tellurium dioxide ≥98.0% (W)

Supplier: TCI America
Warning

T3254-25G
TCT3254-25GEA 73 USD
TCT3254-25G
Tellurium dioxide ≥98.0% (W)
Tellurium dioxide
Formula: O₂Te
Melting Pt: 733°C
MDL Number: MFCD00011263
CAS Number: 7446-07-3
EINECS: 231-193-1
Merck Index: 14,09123

Order Now

Specification Test Results

Appearance White to Orange to Green powder to crystal
Purity (Gravimetric Method) min. 98 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR