α,α,α-Trifluoro-3,5-diaminotoluene ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Danger

T3239-1G T3239-5G
TCT3239-1GEA 40 USD
TCT3239-1G TCT3239-5G
α,α,α-Trifluoro-3,5-diaminotoluene ≥98.0% (by GC, titration analysis)
α,α,α-Trifluoro-3,5-diaminotoluene
Formula: C₇H₇F₃N₂
MDL Number: MFCD00014776
CAS Number: 368-53-6
EINECS: 206-708-8

Order Now

Specification Test Results

Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 87.0 to 91.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR