1,3,5-Tris[(3,3,3-trifluoropropyl)methyl]cyclotrisiloxane ≥98.0% (by GC)

Supplier: TCI America
Danger

Synonyms: 1,3,5-Tris(3,3,3-trifluoropropyl)-1,3,5-trimethylcyclotrisiloxane, 1,3,5-Trimethyl-1,3,5-tris(3,3,3-trifluoropropyl)cyclotrisiloxane

T3226-25G
TCT3226-25GEA 49 USD
TCT3226-25G
1,3,5-Tris[(3,3,3-trifluoropropyl)methyl]cyclotrisiloxane ≥98.0% (by GC)
1,3,5-Tris[(3,3,3-trifluoropropyl)methyl]cyclotrisiloxane
Formula: C₁₂H₂₁F₉O₃Si₃
MDL Number: MFCD00054667
CAS Number: 2374-14-3
EINECS: 219-154-7
UN: 2811
ADR: 6.1,III

Order Now

Specification Test Results

Appearance White to Almost white powder to lump
Purity (GC) min. 98.0 %
Freezing point 22.0 to 26.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR