Tin(II) 2-ethylhexanoate ≥85.0% (by titrimetric analysis)
Supplier: TCI America
|
Warning
|
Synonyms:
2-Ethylcaproic acid tin (II) salt, Tin bis(2-ethylhexanoate), Stannous 2-ethylhexanoate, 2-Ethylhexanoic acid tin (II) salt
T3149-100G
T3149-500G
TCT3149-100GEA
36
USD
TCT3149-100G
TCT3149-500G
Tin(II) 2-ethylhexanoate ≥85.0% (by titrimetric analysis)
Tin (II) 2-ethylhexanoate
Formula:
C₁₆H₃₀O₄Sn |
MDL Number:
MFCD00002676 CAS Number: 301-10-0 EINECS: 206-108-6 |
Specification Test Results
Appearance | Light yellow to Yellow to Orange clear liquid |
Purity (Iodometric Titration) | min. 85.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...