Tris(2,2,2-trifluoroethyl)borate ≥95.0% (by titrimetric analysis)
Supplier: TCI America
CAS Number: 659-18-7
MDL Number: MFCD00093768
Molecular Formula: C6H6BF9O3
Molecular Weight: 307.91
Purity/Analysis Method:
Other Product Names / Synonyms: Boric Acid Tris(2,2,2-trifluoroethyl) Ester
Formula:
C₆H₆BF₉O₃ Boiling Pt: 125 °C Density: 1.44 (20 °C) |
MDL Number:
MFCD00093768 CAS Number: 659-18-7 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...