4-Carboxyphenylboronic acid pinacol ester ≥98.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: 4-Carboxybenzeneboronic acid pinacol ester, 4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzoic acid

T2841-1G
TCT2841-1GEA 31 USD
TCT2841-1G
4-Carboxyphenylboronic acid pinacol ester ≥98.0% (by GC, titration analysis)
4-Carboxyphenylboronic acid pinacol ester
Formula: C₁₃H₁₇BO₄
MDL Number: MFCD01863710
CAS Number: 180516-87-4

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 228.0 to 232.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR