2,3,5-Trifluoropyridine ≥98.0%

Supplier: TCI America

T2778-25G T2778-5G
TCT2778-25GEA 296.1 USD
TCT2778-25G TCT2778-5G
2,3,5-Trifluoropyridine ≥98.0%
2,3,5-Trifluoropyridine

CAS Number: 76469-41-5 MDL Number: MFCD03001162 Molecular Formula: C5H2F3N Molecular Weight: 133.07 Purity/Analysis Method: 98.0% (GC) Form: Clear Liquid Color: Colorless Boiling point (°C): 102 Flash Point (°C): 30 Specific Gravity (20/20): 1.40

Formula: C₅H₂F₃N
Boiling Pt: 102 °C
Density: 1.40 (20 °C)
Flash Pt: 30 °C
MDL Number: MFCD03001162
CAS Number: 76469-41-5
UN: 1993
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5ed28881634d168cf4749caff302655848acbeca&vid=5ed28881634d168cf4749caff302655848acbeca&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR