2',3',5'-Tri-O-acetylguanosine ≥98.0% (by HPLC, titration analysis)
Supplier: TCI America
CAS Number: 6979-94-8
MDL Number: MFCD00057054
Molecular Formula: C16H19N5O8
Molecular Weight: 409.36
Purity/Analysis Method:
Formula:
C₁₆H₁₉N₅O₈ Melting Pt: 213 °C |
MDL Number:
MFCD00057054 CAS Number: 6979-94-8 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...