2',3',5'-Tri-O-acetylguanosine ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

T2692-1G T2692-5G
TCT2692-1GEA 170 USD
TCT2692-1G TCT2692-5G
2',3',5'-Tri-O-acetylguanosine ≥98.0% (by HPLC, titration analysis)
2',3',5'-Tri-O-acetylguanosine

CAS Number: 6979-94-8 MDL Number: MFCD00057054 Molecular Formula: C16H19N5O8 Molecular Weight: 409.36 Purity/Analysis Method: 98.0% (HPLC,T) Form: Crystal Color: White Melting point (°C): 213 Specific rotation [a]20/D: -21 deg (C=0.4, EtOH) Storage Temperature: 0°C

Formula: C₁₆H₁₉N₅O₈
Melting Pt: 213 °C
MDL Number: MFCD00057054
CAS Number: 6979-94-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e50118e4d63e3e0fe9b5d94aac3e73c2c5f5598b&vid=e50118e4d63e3e0fe9b5d94aac3e73c2c5f5598b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR