Luteolin ≥98.0% (by HPLC)

Supplier: TCI America

T2682-1G T2682-5G
TCT2682-1GEA 109.82 USD
TCT2682-1G TCT2682-5G
Luteolin ≥98.0% (by HPLC)
Luteolin
Formula: C₁₅H₁₀O₆
Melting Pt: 330 °C
MDL Number: MFCD00017309
CAS Number: 491-70-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9430201f735f862de57758d4d99d135ed5745713&vid=9430201f735f862de57758d4d99d135ed5745713&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR