Luteolin ≥98.0% (by HPLC)
Supplier: TCI America
CAS Number: 491-70-3
MDL Number: MFCD00017309
Molecular Formula: C15H10O6
Molecular Weight: 286.24
Purity/Analysis Method:
Product Categories: Bioscience, Antioxidants, Aromatic Polyketides, Polyphenols (Phytochemicals)
Other Product Names / Synonyms: Luteolin
Formula:
C₁₅H₁₀O₆ Melting Pt: 330 °C |
MDL Number:
MFCD00017309 CAS Number: 491-70-3 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...