Tetrabutylammonium tetrafluoroborate ≥98.0% (by total nitrogen basis)

Supplier: TCI America
Warning

T2648-25G
TCT2648-25GEA 121 USD
TCT2648-25G
Tetrabutylammonium tetrafluoroborate ≥98.0% (by total nitrogen basis)
Tetrabutylammonium tetrafluoroborate

CAS Number: 429-42-5 MDL Number: MFCD00011634 Molecular Formula: C16H36BF4N Molecular Weight: 329.27 Purity/Analysis Method: 98.0% (N) Form: Crystal Color: White Melting point (°C): 160

Formula: C₁₆H₃₆BF₄N
Melting Pt: 160 °C
Storage Temperature: Ambient
MDL Number: MFCD00011634
CAS Number: 429-42-5
EINECS: 207-058-8

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (N)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR