2,4,5-Trimethoxybenzoic acid ≥98.0%
Supplier: TCI America
T2627-25G
T2627-5G
TCT2627-25GEA
54
USD
TCT2627-25G
TCT2627-5G
2,4,5-Trimethoxybenzoic acid ≥98.0%
2,4,5-Trimethoxybenzoic acid
CAS Number: 490-64-2
MDL Number: MFCD00002435
Molecular Formula: C10H12O5
Molecular Weight: 212.20
Purity/Analysis Method:
Formula:
C₁₀H₁₂O₅ MW: 212.2 Boiling Pt: 300 Melting Pt: 142 |
MDL Number:
MFCD00002435 CAS Number: 490-64-2 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...