2,3,4-Trifluorophenylboronic acid ≥97.0% (contains varying amounts of Anhydride)
Supplier: TCI America
Synonyms:
2,3,4-Trifluorobenzeneboronic acid, Dihydroxy(2,3,4-trifluorophenyl)borane, (2,3,4-Trifluorophenyl)metaboric acid
T2574-5G
TCT2574-5GEA
33
USD
TCT2574-5G
2,3,4-Trifluorophenylboronic acid ≥97.0% (contains varying amounts of Anhydride)
2,3,4-Trifluorophenylboronic acid
CAS Number: 226396-32-3
MDL Number: MFCD01863168
Molecular Formula: C6H4BF3O2
Molecular Weight: 175.90
Purity/Analysis Method:
Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks, Boronic Acids & Boronic Acid Derivatives
Other Product Names / Synonyms: 2,3,4-Trifluorobenzeneboronic Acid
Formula:
C₆H₄BF₃O₂ Melting Pt: 235 °C |
MDL Number:
MFCD01863168 CAS Number: 226396-32-3 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...