Tetraethyl-1,4,8,11-tetraazacyclotetradecane-1,4,8,11-tetraacetate ≥97.0%
Supplier: TCI America
Synonyms:
1,4,8,11-Tetraazacyclotetradecane-N,N',N'',N'''-tetraacetic acid, tetraethyl ester, 1,4,8,11-Tetraazacyclotetradecane-1,4,8,11-tetraacetic Acid Tetraethyl Ester
CAS Number: 126320-57-8
Molecular Formula: C26H48N4O8
Molecular Weight: 544.69
Purity/Analysis Method:
Product Categories: Synthetic Organic Chemistry, Ligands, Materials Chemistry, Macrocycles for Host-Guest Chemistry
Other Product Names / Synonyms: 1,4,8,11-Tetraazacyclotetradecane-1,4,8,11-tetraacetic Acid Tetraethyl Ester
Formula:
C₂₆H₄₈N₄O₈ Melting Pt: 87 °C |
MDL Number:
MFCD09263316 CAS Number: 126320-57-8 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...