4-(Trifluoromethoxy)phenyl isocyanate ≥98.0%

Supplier: TCI America

Synonyms: 1-Isocyanato-4-(trifluoromethoxy)benzene

T2487-25G T2487-5G
TCT2487-25GEA 76.3 USD
TCT2487-25G TCT2487-5G
4-(Trifluoromethoxy)phenyl isocyanate ≥98.0%
4-(Trifluoromethoxy)phenyl isocyanate

CAS Number: 35037-73-1 MDL Number: MFCD00035702 Molecular Formula: C8H4F3NO2 Molecular Weight: 203.12 Purity/Analysis Method: 98.0% (GC) Form: Clear Liquid Color: Colorless Boiling point (°C): 78 Flash Point (°C): 69 Specific Gravity (20/20): 1.36


Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks Other Product Names / Synonyms: Isocyanic Acid 4-(Trifluoromethoxy)phenyl Ester

Formula: C₈H₄F₃NO₂
Boiling Pt: 78 °C
Density: 1.36 (20 °C)
Flash Pt: 69 °C
MDL Number: MFCD00035702
CAS Number: 35037-73-1
UN: 2206
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e3cb413cf82fd67c53af4cce049689320f0d3b0f&vid=e3cb413cf82fd67c53af4cce049689320f0d3b0f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR