Tetrahydrofuran, anhydrous ≥99.5% (by GC) stabilized

Supplier: TCI America

T2394-500ML
TCT2394-500MLEA 55.06 USD
TCT2394-500ML
Tetrahydrofuran, anhydrous ≥99.5% (by GC) stabilized
Tetrahydrofuran

(stabilized with BHT)
CAS Number: 109-99-9
MDL Number: MFCD00005356
Molecular Formula: C4H8O
Molecular Weight: 72.11
Purity/Analysis Method: >99.5% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 65
Melting point (°C): -108
Flash Point (°C): -15
Specific Gravity (20/20): 0.89


Product Categories: Synthetic Organic Chemistry, Anhydrous Solvents
Other Product Names / Synonyms: THF

Formula: C₄H₈O
Boiling Pt: 65 °C
Melting Pt: –108 °C
Density: 0.89 (20 °C)
Flash Pt: –15 °C
MDL Number: MFCD00005356
CAS Number: 109-99-9
UN: 2056
ADR: 3,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5878b108d8580b5945e2bc4a80d0feb033ea9423&vid=6aa88254eae0694713270f2c1f873b0d07a716ec&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.5% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR