D-ɑ-Tocopherol acetate ≥96.0%

Supplier: TCI America

Synonyms: Vitamin E Acetate (D-form), (+)-ɑ-Tocopherol Acetate

T2322-25G
TCT2322-25GEA 161.1 USD
TCT2322-25G
D-ɑ-Tocopherol acetate ≥96.0%
D-alpha-Tocopherol acetate

CAS Number: 58-95-7 Molecular Formula: C31H52O3 Molecular Weight: 472.75 Purity/Analysis Method: 96.0% (GC) Form: Clear Liquid Color: Colorless Flash Point (°C): 245 Specific rotation [a]20/D: 24 deg


Product Categories: Bioscience, Antioxidants, Vitamins, Terpenes (Others) Other Product Names / Synonyms: Vitamin E Acetate (D-form)

Formula: C₃₁H₅₂O₃
MW: 472.75 g/mol
Flash Pt: 245 °C
MDL Number: MFCD00072052
CAS Number: 58-95-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=817ea5a455ff25eb72492128bb5ba3d578c65d15&vid=817ea5a455ff25eb72492128bb5ba3d578c65d15&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 96.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR