D-α-Tocopherol ≥97.0%

Supplier: TCI America

Synonyms: (+)-α-Tocopherol, Vitamin E Alcohol, (R)-2,5,7,8-Tetramethyl-2-((4R,8R)-4,8,12-trimethyltridecyl)chroman-6-ol, α-Vitamin E, 3,4-Dihydro-2,5,7,8-tetramethyl-2-(4,8,12-trimethyltridecyl)-2H-benzopyran-6-ol, Vitamin E, D(+)-α-Tocopherol

T2309-25G
TCT2309-25GEA 131.3 USD
TCT2309-25G
D-α-Tocopherol ≥97.0%
D-α-Tocopherol

CAS Number: 59-02-9 Molecular Formula: C29H50O2 Molecular Weight: 430.72 Purity/Analysis Method: 97.0% (GC) Form: Clear Liquid Color: Pale Yellow Melting point (°C): 4 Flash Point (°C): 253 Specific Gravity (20/20): 0.95 Specific rotation [a]20/D: 0.5 deg (C=1, EtOH)


Product Categories: Bioscience, Antioxidants, Vitamins, Terpenes (Others) Other Product Names / Synonyms: Vitamin E (D-form)

Formula: C₂₉H₅₀O₂
MW: 430.71 g/mol
Melting Pt: 4 °C
Density: 0.95 (20 °C)
Flash Pt: 253 °C
MDL Number: MFCD00072045
CAS Number: 59-02-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3b409a499a24dd58cfa80c6aaafc37e26c4b44b6&vid=3b409a499a24dd58cfa80c6aaafc37e26c4b44b6&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR