Mannose triflate ≥98.0%
Supplier: TCI America
Synonyms:
(2S,3S,4S,5R,6R)-6-(Acetoxymethyl)-3-(((trifluoromethyl)sulphonyl)oxy)tetrahydro-2H-pyran-2,4,5-triyl triacetate, 1,3,4,6-Tetra-O-acetyl-2-O-(trifluoromethanesulfonyl)-β-D-mannopyranose, 1,3,4,6-Tetra-O-acetyl-2-O-(trifluoromethanesulphonyl)-β-D-mannopyranose, (2S,3S,4S,5R,6R)-6-(Acetoxymethyl)-3-(((trifluoromethyl)sulfonyl)oxy)tetrahydro-2H-pyran-2,4,5-triyl triacetate, TATM
CAS Number: 92051-23-5
Molecular Formula: C15H19F3O12S
Molecular Weight: 480.36
Purity/Analysis Method:
Product Categories: Synthetic Organic Chemistry, Sulfonic Acid Derivatives, Bioscience, Mannose, O-Substituted Sugars
Other Product Names / Synonyms: TATM
Formula:
C₁₅H₁₉F₃O₁₂S Melting Pt: 122 °C |
MDL Number:
MFCD00012353 CAS Number: 92051-23-5 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...