Trimethyl orthopropionate ≥97.0%

Supplier: TCI America

T2288-25G T2288-5G
TCT2288-25GEA 46 USD
TCT2288-25G TCT2288-5G
Trimethyl orthopropionate ≥97.0%
Trimethyl orthopropionate

CAS Number: 24823-81-2 MDL Number: MFCD00043305 Molecular Formula: C6H14O3 Molecular Weight: 134.18 Purity/Analysis Method: 97.0% (GC) Form: Clear Liquid Color: Colorless Boiling point (°C): 128 Flash Point (°C): 19 Specific Gravity (20/20): 0.95


Other Product Names / Synonyms: Orthopropionic Acid Trimethyl Ester, 1,1,1-Trimethoxypropane, Methyl Orthopropionate

Formula: C₆H₁₄O₃
Boiling Pt: 128 °C
Density: 0.95 (20 °C)
Flash Pt: 19 °C
MDL Number: MFCD00043305
CAS Number: 24823-81-2
UN: 3272
ADR: 3,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b44ab0160c21886125e8632cf51506ed1a79d8a0&vid=b44ab0160c21886125e8632cf51506ed1a79d8a0&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR