5-(Trimethylsilyl)-1,3-cyclopentadiene ≥97.0%

Supplier: TCI America

Synonyms: Trimethylsilylcyclopentadiene (mixture of isomers), (2,4-Cyclopentadien-1-yl)trimethylsilane

T2284-1G
TCT2284-1GEA 50 USD
TCT2284-1G TCT2284-5G
5-(Trimethylsilyl)-1,3-cyclopentadiene ≥97.0%
5-(Trimethylsilyl)-1,3-cyclopentadiene

CAS Number: 3559-74-8 Molecular Formula: C8H14Si Molecular Weight: 138.29 Purity/Analysis Method: 97.0% (GC) Form: Clear Liquid Color: Colorless Boiling point (°C): 140 Melting point (°C): -19 Flash Point (°C): 29 Specific Gravity (20/20): 0.85 Storage Temperature: 0-10°C


Product Categories: Synthetic Organic Chemistry, Silicon Compounds (for Synthesis) Other Product Names / Synonyms: (2,4-Cyclopentadien-1-yl)trimethylsilane

Formula: C₈H₁₄Si
Boiling Pt: 140 °C
Melting Pt: –19 °C
Density: 0.85 (20 °C)
Flash Pt: 29 °C
MDL Number: MFCD00010666
CAS Number: 3559-74-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=522ea61534933755abdb114eca8e48b2982e3d8a&vid=522ea61534933755abdb114eca8e48b2982e3d8a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR