D-ɑ-Tocopherylquinone ≥97.0% (by HPLC)
Supplier: TCI America
Synonyms:
Vitamin E Quinone (D-form)
T2283-500MG
TCT2283-500MGEA
282.5
USD
TCT2283-500MG
D-ɑ-Tocopherylquinone ≥97.0% (by HPLC)
D-alpha-Tocopherylquinone
CAS Number: 7559-04-8
Molecular Formula: C29H50O3
Molecular Weight: 446.72
Purity/Analysis Method:
Product Categories: Bioscience, Vitamins, Terpenes (Others)
Other Product Names / Synonyms: Vitamin E Quinone (D-form)
Formula:
C₂₉H₅₀O₃ MW: 446.72 g/mol |
MDL Number:
MFCD00135657 CAS Number: 7559-04-8 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...