N,N,N',N'-Tetrakis(p-tolyl)benzidine ≥98.0% (by HPLC, total nitrogen)
Supplier: TCI America
Synonyms:
N4,N4,N4',N4'-Tetra-p-tolyl-[1,1'-biphenyl]-4,4'-diamine, N,N,N',N'-Tetrakis(4-methylphenyl)benzidine
T2269-5G
TCT2269-5GEA
93
USD
TCT2269-5G
N,N,N',N'-Tetrakis(p-tolyl)benzidine ≥98.0% (by HPLC, total nitrogen)
N,N,N',N'-Tetrakis(p-tolyl)benzidine
CAS Number: 76185-65-4
MDL Number: MFCD00799300
Molecular Formula: C40H36N2
Molecular Weight: 544.74
Purity/Analysis Method:
Product Categories: Materials Chemistry, Organic Light-Emitting Diode (OLED) Materials
Other Product Names / Synonyms: N,N,N',N'-Tetrakis(4-methylphenyl)benzidine
Formula:
C₄₀H₃₆N₂ MW: 544.74 g/mol Melting Pt: 218 °C |
MDL Number:
MFCD00799300 CAS Number: 76185-65-4 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...