Dimethylthexylchlorosilane ≥95.0% (by GC)

Supplier: TCI America

T2116-5G
TCT2116-25GEA 270.89 USD
TCT2116-25G TCT2116-5G
Dimethylthexylchlorosilane ≥95.0% (by GC)
Dimethylthexylchlorosilane

CAS Number: 67373-56-2 MDL Number: MFCD00012175 Molecular Formula: C8H19ClSi Molecular Weight: 178.78 Purity/Analysis Method: 95.0% (GC) Form: Clear Liquid Color: Colorless Boiling point (°C): 56 Flash Point (°C): 46 Specific Gravity (20/20): 0.91


Product Categories: Synthetic Organic Chemistry, Protection & Derivatization Reagents (for Synthesis), Silicon Compounds (for Synthesis), Materials Chemistry, Silane Coupling Agents Other Product Names / Synonyms: Chlorodimethyl(1,1,2-trimethylpropyl)silane, Dimethylthexylsilyl Chloride

Formula: C₈H₁₉ClSi
Boiling Pt: 56 °C
Density: 0.91 (20 °C)
Flash Pt: 46 °C
MDL Number: MFCD00012175
CAS Number: 67373-56-2
UN: 2986
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9c1f477a3ff248f01802e2f198bd06960327b488&vid=9c1f477a3ff248f01802e2f198bd06960327b488&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR