3,4,6-Tri-O-benzyl-2-deoxy-D-glucopyranose ≥98.0%
Supplier: TCI America
T1933-100MG
TCT1933-100MGEA
90
USD
TCT1933-100MG
3,4,6-Tri-O-benzyl-2-deoxy-D-glucopyranose ≥98.0%
3,4,6-Tri-O-benzyl-2-deoxy-D-glucopyranose
CAS Number: 132732-60-6
MDL Number: MFCD06797170
Molecular Formula: C27H30O5
Molecular Weight: 434.53
Purity/Analysis Method:
Formula:
C₂₇H₃₀O₅ |
MDL Number:
MFCD06797170 CAS Number: 132732-60-6 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...