3,4',5-Trimethoxy-trans-stilbene ≥96.0%
Supplier: TCI America
Synonyms:
trans-1-(3,5-Dimethoxyphenyl)-2-(4-methoxyphenyl)ethylene, (E)-1,3-Dimethoxy-5-(4-methoxystyryl)benzene, trans-Trimethoxyresveratrol
T1829-5G
TCT1829-005GEA
438.2
USD
TCT1829-005G
3,4',5-Trimethoxy-trans-stilbene ≥96.0%
3,4',5-Trimethoxy-trans-stilbene
CAS Number: 22255-22-7
MDL Number: MFCD02093500
Molecular Formula: C17H18O3
Molecular Weight: 270.33
Purity/Analysis Method:
Product Categories: Bioscience, Aromatic Polyketides
Other Product Names / Synonyms: trans-1-(3,5-Dimethoxyphenyl)-2-(4-methoxyphenyl)ethylene
Formula:
C₁₇H₁₈O₃ MW: 270.33 g/mol Melting Pt: 57 °C |
MDL Number:
MFCD02093500 CAS Number: 22255-22-7 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...