L-Threitol ≥98.0%
Supplier: TCI America
Synonyms:
L-1,2,3,4-Butanetetrol
T1647-1G
TCT1647-001GEA
158
USD
TCT1647-001G
L-Threitol ≥98.0%
L-Threitol
CAS Number: 2319-57-5
MDL Number: MFCD00064294
Molecular Formula: C4H10O4
Molecular Weight: 122.12
Purity/Analysis Method:
Product Categories: Bioscience, Sugar Alcohols, Sugars (Others)
Other Product Names / Synonyms: L-1,2,3,4-Butanetetrol
Formula:
C₄H₁₀O₄ MW: 122.12 g/mol Melting Pt: 90 °C |
MDL Number:
MFCD00064294 CAS Number: 2319-57-5 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...