5,10,15,20-Tetrakis(2,6-dichlorophenyl)porphyrin ≥95.0% (by HPLC)
Supplier: TCI America
Synonyms:
5,10,15,20-Tetrakis(2,6-dichlorophenyl)porphine
T1438-100MG
TCT1438-100MGEA
221.8
USD
TCT1438-100MG
5,10,15,20-Tetrakis(2,6-dichlorophenyl)porphyrin ≥95.0% (by HPLC)
5,10,15,20-Tetrakis(2,6-dichlorophenyl)porphyrin
CAS Number: 37083-37-7
MDL Number: MFCD00191500
Molecular Formula: C44H22Cl8N4
Molecular Weight: 890.29
Purity/Analysis Method:
Product Categories: Synthetic Organic Chemistry, Oxidation, Ligands, Materials Chemistry, Porphyrin & Phthalocyanine Dyes (Functional Organic Dyes), Bioscience, Porphyrins
Other Product Names / Synonyms: 5,10,15,20-Tetrakis(2,6-dichlorophenyl)porphine
Formula:
C₄₄H₂₂Cl₈N₄ MW: 890.29 g/mol |
MDL Number:
MFCD00191500 CAS Number: 37083-37-7 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...