Trigonelline hydrochloride ≥98.0% (by HPLC, titration analysis)
Supplier: TCI America
CAS Number: 6138-41-6
MDL Number: MFCD00077250
Molecular Formula: C7H8ClNO2
Molecular Weight: 173.60
Purity/Analysis Method:
Product Categories: Bioscience, Crude Medicine Ingredients for Research and Experimental Use, Pyridine Alkaloids
Other Product Names / Synonyms: N-Methylnicotinic Acid Betaine Hydrochloride, 1-Methylpyridinium-3-carboxylate Hydrochloride
Formula:
C₇H₈ClNO₂ |
MDL Number:
MFCD00077250 CAS Number: 6138-41-6 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...