1,4,7,10-Tetraazacyclododecane tetrahydrochloride ≥98.0%

Supplier: TCI America

Synonyms: Cyclen tetrahydrochloride

T1426-5G T1426-1G
TCT1426-005GEA 333.7 USD
TCT1426-005G TCT1426-001G
1,4,7,10-Tetraazacyclododecane tetrahydrochloride ≥98.0%
1,4,7,10-Tetraazacyclododecane tetrahydrochloride

CAS Number: 10045-25-7 MDL Number: MFCD00068484 Molecular Formula: C8H20N4 Molecular Weight: 318.11 Purity/Analysis Method: 98.0% (N) Form: Crystal Color: White Melting point (°C): 280


Product Categories: Synthetic Organic Chemistry, Ligands, Materials Chemistry, Macrocycles for Host-Guest Chemistry, Bioscience, Polyamines, Analytical Science, Chelating Agents for Magnetic Resonance Imaging (MRI) Other Product Names / Synonyms: Cyclen-4HCl

Formula: C₈H₂₄Cl₄N₄
MW: 318.12 g/mol
Melting Pt: 280 °C
MDL Number: MFCD00068484
CAS Number: 10045-25-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ae7cfbd6d16e119459911c8c54fc63bb262bdef2&vid=ae7cfbd6d16e119459911c8c54fc63bb262bdef2&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (N)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR