1,4,7,10-Tetraazacyclododecane tetrahydrochloride ≥98.0%
Supplier: TCI America
Synonyms:
Cyclen tetrahydrochloride
T1426-5G
T1426-1G
TCT1426-005GEA
333.7
USD
TCT1426-005G
TCT1426-001G
1,4,7,10-Tetraazacyclododecane tetrahydrochloride ≥98.0%
1,4,7,10-Tetraazacyclododecane tetrahydrochloride
CAS Number: 10045-25-7
MDL Number: MFCD00068484
Molecular Formula: C8H20N4
Molecular Weight: 318.11
Purity/Analysis Method:
Product Categories: Synthetic Organic Chemistry, Ligands, Materials Chemistry, Macrocycles for Host-Guest Chemistry, Bioscience, Polyamines, Analytical Science, Chelating Agents for Magnetic Resonance Imaging (MRI)
Other Product Names / Synonyms: Cyclen-4HCl
Formula:
C₈H₂₄Cl₄N₄ MW: 318.12 g/mol Melting Pt: 280 °C |
MDL Number:
MFCD00068484 CAS Number: 10045-25-7 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...