2,2,2-Tribromoethanol ≥98.0%

Supplier: TCI America
Warning

Synonyms: Tribromoethyl alcohol

T1420-25G
TCT1420-025GEA 119.02 USD
TCT1420-025G
2,2,2-Tribromoethanol ≥98.0%
2,2,2-Tribromoethanol

CAS Number: 75-80-9 MDL Number: MFCD00004671 Molecular Formula: C2H3Br3O Molecular Weight: 282.76 Purity/Analysis Method: 98.0% (GC) Form: Crystal Color: White Boiling point (°C): 94 Melting point (°C): 81 Flash Point (°C): 110

Formula: Br₃CCH₂OH
Boiling Pt: 94 °C
Melting Pt: 81 °C
Flash Pt: 110 °C
Storage Temperature: Ambient
MDL Number: MFCD00004671
CAS Number: 75-80-9
EINECS: 200-903-1
Merck Index: 12,09743
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=73970c813c0d84770c447b5b10bff41d439cb48c&vid=73970c813c0d84770c447b5b10bff41d439cb48c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR