1-(p-Toluenesulfonyl)pyrrole ≥98.0% (by titrimetric analysis)
Supplier: TCI America
CAS Number: 17639-64-4
MDL Number: MFCD00145014
Molecular Formula: C11H11NO2S
Molecular Weight: 221.27
Purity/Analysis Method:
Product Categories: Materials Chemistry, Conducting Polymer Research Reagents
Other Product Names / Synonyms: 1-Tosylpyrrole
Formula:
C₁₁H₁₁NO₂S Melting Pt: 103 °C |
MDL Number:
MFCD00145014 CAS Number: 17639-64-4 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...