meso-Tetraphenylporphine ≥98.0% (by HPLC) chlorin-free

Supplier: TCI America

Synonyms: 5,10,15,20-Tetraphenyl-21H,23H-porphine, 5,10,15,20-Tetraphenylporphyrin, meso-Tetraphenylporphine, TPP

T1359-1G
TCT1359-001GEA 179 USD
TCT1359-001G
meso-Tetraphenylporphine ≥98.0% (by HPLC) chlorin-free
meso-Tetraphenylporphine

CAS Number: 917-23-7 MDL Number: MFCD00011680 Molecular Formula: C44H30N4 Molecular Weight: 614.75 Purity/Analysis Method: 98.0% (HPLC) Form: Crystal Color: Deep Red


Product Categories: Synthetic Organic Chemistry, Oxidation, Ligands, Materials Chemistry, Precise Polymerization Reagents, Porphyrin & Phthalocyanine Dyes (Functional Organic Dyes), Bioscience, Porphyrins Other Product Names / Synonyms: Tetraphenylporphine, TPP

Formula: C₄₄H₃₀N₄
MW: 614.75 g/mol
Density: 1.276 g/cm³ (20 °C)
MDL Number: MFCD00011680
CAS Number: 917-23-7
EINECS: 213-025-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b1d61bb737987a76c7c53f2ddc02a66eca906497&vid=b1d61bb737987a76c7c53f2ddc02a66eca906497&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR