4-Trifluoroacetyl-3-methyl-1-phenyl-5-pyrazolone ≥96.0% (by titrimetric analysis)
Supplier: TCI America
CAS Number: 1691-93-6
MDL Number: MFCD00143378
Molecular Formula: C12H9F3N2O2
Molecular Weight: 270.21
Purity/Analysis Method:
Other Product Names / Synonyms: 3-Methyl-1-phenyl-4-trifluoroacetyl-5-pyrazolone
Formula:
C₁₂H₉F₃N₂O₂ Melting Pt: 147 °C |
MDL Number:
MFCD00143378 CAS Number: 1691-93-6 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...