(3-Aminopropyl)trimethoxysilane ≥96.0%

Supplier: TCI America
Warning

Synonyms: 3-(Trimethoxysilyl)propylamine, 3-(Trimethoxysilyl)propan-1-amine, Trimethoxy(3-aminopropyl)silane, 3-Aminopropyltrimethoxysilane

T1255-500ML T1255-25ML T1255-100ML
TCT1255-500MLEA 136 USD
TCT1255-500ML TCT1255-25ML TCT1255-100ML
(3-Aminopropyl)trimethoxysilane ≥96.0%
(3-Aminopropyl)trimethoxysilane

CAS Number: 13822-56-5 MDL Number: MFCD00008206 Molecular Formula: C6H17NO3Si Molecular Weight: 179.29 Purity/Analysis Method: 96.0% (GC,T) Form: Clear Liquid Color: Colorless Boiling point (°C): 92 Flash Point (°C): 86 Specific Gravity (20/20): 1.02


Product Categories: Materials Chemistry, Silane Coupling Agents Other Product Names / Synonyms: 3-(Trimethoxysilyl)propylamine

Formula: H₂NCH₂CH₂CH₂Si(OCH₃)₃
MW: 179.29 g/mol
Boiling Pt: 194
Density: 1.02 (20 °C)
Flash Pt: 86 °C
Storage Temperature: Ambient
MDL Number: MFCD00008206
CAS Number: 13822-56-5
EINECS: 237-511-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8c41d614ba064bf2c6322083519a165138bb684c&vid=8c41d614ba064bf2c6322083519a165138bb684c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 96.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR