Tetramethylammonium hydroxide 10% in methanol

Supplier: TCI America

T1201-25ML
TCT1201-025MLEA 94.24 USD
TCT1201-025ML
Tetramethylammonium hydroxide 10% in methanol
Tetramethylammonium hydroxide

[for Photoresist Research]
CAS Number: 75-59-2
MDL Number: MFCD00008280
Molecular Formula: C4H13NO
Molecular Weight: 91.15
Form: Clear Liquid
Color: Colorless
Flash Point (°C): 12
Specific Gravity (20/20): 0.83


Other Product Names / Synonyms: TMAH

Formula: C₄H₁₃NO
MW: 91.15 g/mol
Density: 0.83 (20 °C)
Flash Pt: 12 °C
Storage Temperature: Ambient
MDL Number: MFCD00008280
CAS Number: 75-59-2
EINECS: 200-882-9
UN: 1835
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5be4dc74437d365abce27792066b00c181fba48f&vid=3beb55b8a2eaa0a73ff553dfc3ccc34a830e648b&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR