Tin (II) trifluoromethanesulfonate ≥90.0%

Supplier: TCI America

Synonyms: Tin trifluoromethanesulfonate, Tin(II) trifluoromethanesulfonate, Tin trifluoromethanesulphonate, Tin triflate, Tin(II) trifluoromethanesulphonate

T1194-1G T1194-5G T1194-25G
TCT1194-001GEA 17 USD
TCT1194-001G TCT1194-005G TCT1194-25G
Tin (II) trifluoromethanesulfonate ≥90.0%
Tin (II) trifluoromethanesulphonate

CAS Number: 62086-04-8 MDL Number: MFCD00191251 Molecular Formula: CHF3O3S Molecular Weight: 416.84 Purity/Analysis Method: 90.0% (T) Form: Crystal Color: White


Product Categories: Synthetic Organic Chemistry, Homogeneous Catalysts, Homogeneous Catalysts Other Product Names / Synonyms: Stannous Trifluoromethanesulfonate, Tin(II) Triflate, Trifluoromethanesulfonic Acid Tin(II) Salt

Formula: C₂F₆O₆S₂Sn
MDL Number: MFCD00191251
CAS Number: 62086-04-8
UN: 3288
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=612c224942d6c4076b341dc417b10216899c437b&vid=612c224942d6c4076b341dc417b10216899c437b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 90.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR