Tricyclohexylphosphine ∼18% ∼0.60 mol/L in toluene (contains Tricyclohexylphosphine Oxide)
Supplier: TCI America
T1165-25ML
TCT1165-25MLEA
69
USD
TCT1165-25ML
Tricyclohexylphosphine ∼18% ∼0.60 mol/L in toluene (contains Tricyclohexylphosphine Oxide)
Tricyclohexylphosphine
CAS Number: 2622-14-2
MDL Number: MFCD00003853
Molecular Formula: C18H33P
Molecular Weight: 280.44
Form: Clear Liquid
Color: Colorless
Flash Point (°C): 42
Formula:
C₁₈H₃₃P Flash Pt: 42 °C |
MDL Number:
MFCD00003853 CAS Number: 2622-14-2 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...