Tetrabutylammonium fluoride ∼1 mol/L in THF

Supplier: TCI America

T1125-100ML T1125-25ML
TCT1125-100MLEA 161.1 USD
TCT1125-100ML TCT1125-25ML
Tetrabutylammonium fluoride ∼1 mol/L in THF
Tetrabutylammonium fluoride

[for Catalyst of acylation, silylation and cleavage of Silyl Ether]
CAS Number: 429-41-4
MDL Number: MFCD00011747
Molecular Formula: C16H36FN
Molecular Weight: 261.47
Form: Clear Liquid
Color: Very Pale Yellow
Flash Point (°C): -17
Specific Gravity (20/20): 0.91


Other Product Names / Synonyms: TBAF

Formula: C₁₆H₃₆FN
MW: 261.47
Density: 0.91 (20 °C)
Flash Pt: –17 °C
MDL Number: MFCD00011747
CAS Number: 429-41-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9ea3e7d20f436301c6826b3631daf8f303830a26&vid=53a3129f4faa84a600009e09b936103ffc97e7d8&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR