1-(Trimethylsilyl)-1-propyne ≥98.0%

Supplier: TCI America

Synonyms: Trimethyl-1-propynylsilane

T1123-100ML T1123-25ML T1123-5ML
TCT1123-100MLEA 635 USD
TCT1123-100ML TCT1123-25ML TCT1123-5ML
1-(Trimethylsilyl)-1-propyne ≥98.0%
1-(Trimethylsilyl)-1-propyne

CAS Number: 6224-91-5 MDL Number: MFCD00009271 Molecular Formula: C6H12Si Molecular Weight: 112.25 Purity/Analysis Method: 98.0% (GC) Form: Clear Liquid Color: Colorless Boiling point (°C): 100 Flash Point (°C): -4 Specific Gravity (20/20): 0.75 Storage Temperature: 0-10°C


Product Categories: Synthetic Organic Chemistry, Silicon Compounds (for Synthesis), Materials Chemistry, Langmuir-Blodgett(LB) Film Forming Agents Other Product Names / Synonyms: 1-TMS-1-propyne

Formula: C₆H₁₂Si
MW: 112.25
Boiling Pt: 99
Density: 0.75 (20 °C)
Flash Pt: –4 °C
MDL Number: MFCD00009271
CAS Number: 6224-91-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6e68fc5abc6fa38b17cb3ee204d930a8ee8069e7&vid=6e68fc5abc6fa38b17cb3ee204d930a8ee8069e7&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR