Tetraethylammonium tetrafluoroborate ≥98.0%

Supplier: TCI America

Synonyms: Tetrylammonium tetrafluoroborate

T0983-25G
TCT0983-25GEA 69 USD
TCT0983-25G
Tetraethylammonium tetrafluoroborate ≥98.0%
Tetraethylammonium tetrafluoroborate

CAS Number: 429-06-1 MDL Number: MFCD00011827 Molecular Formula: C8H20BF4N Molecular Weight: 217.06 Purity/Analysis Method: 98.0% (N) Form: Crystal Color: White

Formula: C₈H₂₀BF₄N
MDL Number: MFCD00011827
CAS Number: 429-06-1
UN: 2811
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=59e6d16dca315c620a6a158a252a38b7e4180a90&vid=59e6d16dca315c620a6a158a252a38b7e4180a90&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (N)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR