Tetrabutylammonium hydrogen sulfate ≥98.0%
Supplier: TCI America
|
Warning
|
Synonyms:
Tetrabutylammonium bisulphate, Tetrabutylammonium hydrogensulfate, Tetrabutylammonium bisulfate, Tetrabutylammonium hydrogensulphate
T0835-25G
T0835-500G
T0835-100G
TCT0835-025GEA
29.52
USD
TCT0835-025G
TCT0835-500G
TCT0835-100G
Tetrabutylammonium hydrogen sulfate ≥98.0%
Tetrabutylammonium hydrogen sulphate
CAS Number: 32503-27-8
MDL Number: MFCD00011637
Molecular Formula: C16H37NO4S
Molecular Weight: 339.54
Purity/Analysis Method:
Formula:
(C₄H₉)₄NHSO₄ Melting Pt: 173 °C Density: 1.01 g/cm³ (25 °C) |
MDL Number:
MFCD00011637 CAS Number: 32503-27-8 EINECS: 251-068-5 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...