TMS-PZ (=N-trimethylsilylimidazole in anhydrous pyridine) kit silylation reagent

Supplier: TCI America
Danger

Synonyms: N-Trimethylsilylimidazole in anhydrous pyridine, SIM in anhydrous pyridine

T0692-1KIT
TCT0692-KITEA 151.7 USD
TCT0692-KIT
TMS-PZ (=N-trimethylsilylimidazole in anhydrous pyridine) kit silylation reagent
TMS-PZ (=N-trimethylsilylimidazole in anhydrous pyridine)

N-(Trimethylsilyl)-1H-imidazole in Anhydrous Pyridine. Trimethylsilylating Reagent, for wet or dry Sugars, Mono or Poly Saccharides.


Kit contains 1 mL × 8 / Reaction vial, capacity 2 mL × 8

Storage Temperature: Refrigerator
MDL Number: MFCD00148688
UN: 2924
ADR: 3.8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR