N,O-Bis(trimethylsilyl)acetamide kit 25% in acetonitrile trimethylsilylating reagent, for NH₂ compounds

Supplier: TCI America
Danger

Synonyms: BSA, Silyl Derivatisation, Trimethylsilyl N-(trimethylsilyl)acetimidate, Trimethylsilyl N-trimethylsilylacetamidate

T0691-1KIT
TCT0691-1KITEA 92.7 USD
TCT0691-1KIT
N,O-Bis(trimethylsilyl)acetamide kit 25% in acetonitrile trimethylsilylating reagent, for NH₂ compounds
N,O-Bis(trimethylsilyl)acetamide

N,O-Bis(trimethylsilyl)acetamide Kit TMS-BA (25% in Acetonitrile) 1 mL × 8 / Reaction vial, capacity 2 mL × 8 [Trimethylsilylating Reagent, for NH2 compounds]

Formula: C₈H₂₁NOSi₂
MW: 203.43 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00008270
CAS Number: 10416-59-8
EINECS: 233-892-7
UN: 1992
ADR: 3,II

Order Now

Specification Test Results

Appearance Colorless to Light yellow clear liquid
Effect test of TMS derivatization Effective as derivatization agent DL-Threonine

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR