n-Tetratetracontane ≥97.0%

Supplier: TCI America

T0629-100MG T0629-1G
TCT0629-100MGEA 35 USD
TCT0629-100MG TCT0629-001G
n-Tetratetracontane ≥97.0%
n-Tetratetracontane

CAS Number: 7098-22-8 MDL Number: MFCD00015268 Molecular Formula: C44H90 Molecular Weight: 619.20 Purity/Analysis Method: 97.0% (GC) Form: Crystal Color: White Melting point (°C): 86 Flash Point (°C): 93

Formula: H₃C(CH₂)₄₂CH₃
MW: 619.2 g/mol
Melting Pt: 86 °C
Flash Pt: 93 °C
Storage Temperature: Ambient
MDL Number: MFCD00015268
CAS Number: 7098-22-8
EINECS: 230-407-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2bcd8e3bfde7b73c74ee0b497228f77167682b9b&vid=788bcca572193a58844c5121878cbbc8c1f65430&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR