TMS-PZ (=N-trimethylsilylimidazole in anhydrous pyridine) silylation reagent

Supplier: TCI America
Danger

Synonyms: N-Trimethylsilylimidazole in anhydrous pyridine, SIM in anhydrous pyridine

T0623-12ML
TCT0623-12MLEA 138 USD
TCT0623-12ML
TMS-PZ (=N-trimethylsilylimidazole in anhydrous pyridine) silylation reagent
TMS-PZ (=N-trimethylsilylimidazole in anhydrous pyridine)

Trimethylsilylating Reagent, for wet or dry Sugars, Mono or Poly Saccharides


Product Categories: Analytical Science, Silylation (GC Derivatizing Reagents)

Flash Pt: 6 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00148688
UN: 2924
ADR: 3,II

Order Now

Specification Test Results

Appearance Colorless to Almost colorless clear liquid
Effect test of TMS derivatization Effective as derivatization agent D-(+)-Glucose
High boiling impurity of Silylation agent No impurity higher than Isopropyl myristate(50 ppm) after 5 min + hold up time

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR