Chlorotriethylsilane ≥97.0%

Supplier: TCI America

Synonyms: Triethylchlorosilane

T0589-5G T0589-25G T0589-100G
TCT0589-005GEA 23.88 USD
TCT0589-005G TCT0589-025G TCT0589-100G
Chlorotriethylsilane ≥97.0%
Chlorotriethylsilane

CAS Number: 994-30-9 MDL Number: MFCD00000507 Molecular Formula: C6H15ClSi Molecular Weight: 150.72 Purity/Analysis Method: 97.0% (GC) Form: Clear Liquid Color: Colorless Boiling point (°C): 149 Flash Point (°C): 30 Specific Gravity (20/20): 0.90


Product Categories: Synthetic Organic Chemistry, Protection & Derivatization Reagents (for Synthesis), Materials Chemistry, Silane Coupling Agents Other Product Names / Synonyms: Triethylsilyl Chloride, Triethylchlorosilane, TES-Cl

Formula: C₆H₁₅ClSi
Boiling Pt: 149 °C
Density: 0.90 (20 °C)
Flash Pt: 30 °C
Storage Temperature: Ambient
MDL Number: MFCD00000507
CAS Number: 994-30-9
EINECS: 213-615-6
UN: 2986
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c2f49726370a1e3d79849dcdcbd1037226158148&vid=c2f49726370a1e3d79849dcdcbd1037226158148&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR